missing translation for 'onlineSavingsMsg'
Learn More

Tracazolate hydrochloride, Tocris Bioscience™

Allosteric modulator of GABAA receptors

Brand:  Tocris (Bio-Techne) 1558/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13469673

  • £308.70 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GABAA receptor modulator. Can potentiate or inhibit recombinant GABAA function depending on subunit combination. Enhances native GABAA receptor function and is anxiolytic following systemic administration in vivo.
Specifications

Chemical Identifiers

1135210-68-2
340.852
tracazolate hydrochloride, dsstox_cid_25208, dsstox_rid_80751, dsstox_gsid_45208, cas-nocas_45208, 4-butylamino-1-ethyl-6-methyl-1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester
ethyl 4-(butylamino)-1-ethyl-6-methylpyrazolo[3,4-b]pyridine-5-carboxylate;hydrochloride
C16H25ClN4O2
TUIXDPUEMQXVHO-UHFFFAOYSA-N
24871267
CCCCNC1=C2C=NN(C2=NC(=C1C(=O)OCC)C)CC.Cl

Specifications

50mg
1135210-68-2
tracazolate hydrochloride, dsstox_cid_25208, dsstox_rid_80751, dsstox_gsid_45208, cas-nocas_45208, 4-butylamino-1-ethyl-6-methyl-1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester
CCCCNC1=C2C=NN(C2=NC(=C1C(=O)OCC)C)CC.Cl
340.852
340.85
Tracazolate hydrochloride
C16H25ClN4O2
TUIXDPUEMQXVHO-UHFFFAOYSA-N
ethyl 4-(butylamino)-1-ethyl-6-methylpyrazolo[3,4-b]pyridine-5-carboxylate;hydrochloride
24871267
>99%
Videos
Documents