missing translation for 'onlineSavingsMsg'
Learn More

TRA-1-81 (Podocalyxin), PE, clone: TRA-1-81, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-8883-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299329

  • £62.71 / 25µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The TRA-1-81 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embyronal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-81 antibody is lost upon cell differentiation. The TRA-1-81 epitope is resistant to neuraminidase digestion, unlike the epitope recognized by the related TRA-1-60 antibody. The TRA-1-81 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer.Applications Reported: This TRA-1-81 antibody has been reported for use in flow cytometric analysis.Applications Tested: This TRA-1-81 antibody has been tested by flow cytometric analysis of the human embryonal carcinoma (EC) line 2102Ep. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TRA-1-81 (podocalyxin)
TRA-1-81
PE
Conjugated
IgM
RUO
Primary
Human
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos