Learn More
TRA-1-60 (Podocalyxin) Monoclonal Antibody (TRA-1-60), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-8863-82
77.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The TRA-1-60 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embryonal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-60 antibody is lost upon cell differentiation. Contrary to early reports that the TRA-1-60 epitope can be destroyed by neuraminidase digestion, new strudies have shown that TRA-1-60 recognizes a neurominidase resistent antigen. The TRA-1-60 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer. Applications Reported: This TRA-1-60 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TRA-1-60 antibody has been tested by flow cytometric analysis of the human embryonal carcinoma (EC) line 2102Ep. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.Specifications
TRA-1-60 (Podocalyxin) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |
Flow Cytometry | |
TRA-1-60 | |
Biotin | |
TRA160; tra60 | |
Affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgM |
For Research Use Only.