missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557725
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SLESEGAGEG AAASPELSAL EEAFRRFAVH GDARATGREM HGKNWSKLC Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
TPPP Polyclonal antibody specifically detects TPPP in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TPPP | |
Polyclonal | |
Unconjugated | |
TPPP | |
25 kDa brain-specific protein; 2900041A09Rik; AI849835; brain specific protein p25 alpha; brain-specific protein p25 alpha; glycogen synthase kinase 3 (GSK3) inhibitor p24; p24; p25; p25alpha; p25-alpha; RGD1310121; Tppp; TPPP/p25; TPPP1; tubulin polymerization promoting protein; tubulin polymerization-promoting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11076 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94811 | |
TPPP | |
Recombinant protein corresponding to Human TPPP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only