missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ TPE (TRIS + Phosphate + EDTA), 10x

Brand:  Alfa Aesar™ J60233.K3

Code : NEW

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 15403039

  • £33.35 / 2 L

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Buffer
Colorless
2L
10X
Liquid
Liquid
Videos
SDS Documents
Certificates