missing translation for 'onlineSavingsMsg'
Learn More

TOX, PE, clone: TXRX10, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-6502-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209259

  • £168.34 / 100µg


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This TXRX10 monoclonal antibody reacts with human and mouse TOX, which is also known as Thymocyte selection-associated HMG box protein. This DNA-binding nuclear factor is expressed in thymocytes, T lymphocytes, NK cells, and lymphoid tissue-inducer (LTi) cells. TOX expression in thymocytes is upregulated during beta-selection and positive selection. Studies using Tox knockout mice have demonstrated a critical role for TOX in CD4+ T cell development as a regulator of the CD4loCD8lo to CD4+CD8lo transition. Moreover, TOX has been shown to be involved in the development of NK and LTi cells. While its regulation remains unclear, TOX expression has been shown to be induced by calcineurin-mediated TCR signaling in thymocytes during positive selection.Applications Reported: This TXRX10 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This TXRX10 antibody has been tested intracellular flow cytometric analysis of mouse thymocytes using the Foxp3 Buffers (cat. 00-5521) and protocol. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TOX
TXRX10
PE
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Thymocyte selection-associated HMG box protein
IgG2a
RUO
Primary
Murine
Videos