missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves

Product Code. 15201588 Shop All Ansell Products
Click to view available options
Size:
9.5
Large
Medium
Small
X-Large
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 15201588

Brand: Ansell™ 93300_9.510

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Complies with ASTM D 3577, Type II. Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves are ideal for use in electronics and semiconductor manufacturing, and laboratory testing.

  • Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove
  • Provides the wearer with a high level of comfort and increased protection in the forearm (longer cuff)
  • Manufactured from nitrile, offering three times the puncture resistance of standard latex gloves
  • Made without natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type l) wearers
  • Suitable for ISO Class 5 or higher clean room environments
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nitrile
Certifications/Compliance Standards CATEGORY III EN 374, EN 421, ASTM D 3577, Type II
Cleanroom Class Class 100 (ISO 5)
Cuff Material Nitrile
Accelerator Free No
AQL 1.5
Coating Material Nitrile
Color Green
Disposable Single-use
Exterior Finish Fingertip Textured
Hand Specific
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 9.5
Sterility Non-sterile
Thickness (Metric) 0.12 mm
Finger Style Textured fingertips
For Use With (Application) For blending of liquids, filtration processes, vaccines injectables operations etc.
Quantity 100 Pairs
Resistant To Splash and high puncture resistance
Series 93-300
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.