missing translation for 'onlineSavingsMsg'
Learn More

Torcetrapib, Tocris Bioscience™

Inhibitor of cholesteryl ester transfer protein (CETP)

Brand:  Tocris (Bio-Techne) 418450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13434304

  • £633.57 / 50mg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of cholesteryl ester transfer protein (CETP). Also impairs endothelial function in vivo; induces hypertension and inhibits acetylcholine-induced vasodilation in rabbit central ear artery.
Specifications

Chemical Identifiers

262352-17-0
600.482
torcetrapib, torcetrapib usan, --torcetrapib, unii-4n4457mv2u, torcetropib, 2r,4s-4-3,5-bis-trifluoromethylbenzyl methoxycarbonylamino-2-ethyl-6-trifluoromethyl-3,4-dihydro-2h-quinoline-1-carboxylic acid ethyl ester, torcetrapib usan/inn, ethyl 2r,4s-4-3,5-bis trifluoromethyl benzyl methoxycarbonyl amino-2-ethyl-6-trifluoromethyl-3,4-dihydroquinoline-1 2h-carboxylate
CHEBI:49203
CCC1CC(C2=C(N1C(=O)OCC)C=CC(=C2)C(F)(F)F)N(CC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(=O)OC
C26H25F9N2O4
CMSGWTNRGKRWGS-NQIIRXRSSA-N
159325
ethyl (2R,4S)-4-[[3,5-bis(trifluoromethyl)phenyl]methyl-methoxycarbonylamino]-2-ethyl-6-(trifluoromethyl)-3,4-dihydro-2H-quinoline-1-carboxylate

Specifications

50mg
262352-17-0
torcetrapib, torcetrapib usan, --torcetrapib, unii-4n4457mv2u, torcetropib, 2r,4s-4-3,5-bis-trifluoromethylbenzyl methoxycarbonylamino-2-ethyl-6-trifluoromethyl-3,4-dihydro-2h-quinoline-1-carboxylic acid ethyl ester, torcetrapib usan/inn, ethyl 2r,4s-4-3,5-bis trifluoromethyl benzyl methoxycarbonyl amino-2-ethyl-6-trifluoromethyl-3,4-dihydroquinoline-1 2h-carboxylate
CCC1CC(C2=C(N1C(=O)OCC)C=CC(=C2)C(F)(F)F)N(CC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(=O)OC
600.482
CHEBI:49203
>99%
Torcetrapib
C26H25F9N2O4
CMSGWTNRGKRWGS-NQIIRXRSSA-N
ethyl (2R,4S)-4-[[3,5-bis(trifluoromethyl)phenyl]methyl-methoxycarbonylamino]-2-ethyl-6-(trifluoromethyl)-3,4-dihydro-2H-quinoline-1-carboxylate
159325
600.47
Videos
Documents