Learn More
Topotecan hydrochloride, Tocris Bioscience™
DNA topoisomerase I inhibitor; Camptothecin (Cat. No. 1100) analog
Brand:  RD Systems (Bio-Techne) 4562/50
Code : Z1
Additional Details : Weight : 0.00050kg
Quantity 50mg
 View more versions of this product
Product Code. 13444934
  • £175.56 / 50mg
Estimated Shipment
28-06-2019
Description
Description
DNA topoisomerase I inhibitor; induces rapid apoptotic death in human B-lineage acute lymphoblastic leukemia (ALL) cells. Improves survival in three mouse models of poor prognosis ALL; downregulates B-Myb and MycN expression in neuroblastoma cells. Water-soluble analog of Camptothecin (Cat. No. 1100). Orally bioavailable.
Specifications
Chemical Identifiers
119413-54-6
457.911
topotecan hydrochloride, topotecan hcl, hycamptamine hydrochloride, topotecan hydrochloride, topotecan monohydrochloride, unii-956s425zcy, topotecan hydrochloride usan, nogitecan hydrochloride, dsstox_cid_25952, dsstox_rid_81248
CCC1(C2=C(COC1=O)C(=O)N3CC4=C(C3=C2)N=C5C=CC(=C(C5=C4)CN(C)C)O)O.Cl
C23H24ClN3O5
DGHHQBMTXTWTJV-BQAIUKQQSA-N
60699
Specifications
Topotecan hydrochloride
119413-54-6
topotecan hydrochloride, topotecan hcl, hycamptamine hydrochloride, topotecan hydrochloride, topotecan monohydrochloride, unii-956s425zcy, topotecan hydrochloride usan, nogitecan hydrochloride, dsstox_cid_25952, dsstox_rid_81248
CCC1(C2=C(COC1=O)C(=O)N3CC4=C(C3=C2)N=C5C=CC(=C(C5=C4)CN(C)C)O)O.Cl
60699
>98%
50mg
C23H24ClN3O5
DGHHQBMTXTWTJV-BQAIUKQQSA-N
457.911
457.91
Videos