Learn More
TOMM20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552843
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KRRSDPNFKN RLRERRKKQK LAKERAGLSK LPDLKDAEAV QKFFLEEIQL GEELLAQGEY EKGVDHLTNA IAVCGQPQQL LQVLQQTLPP PVFQMLLTKL PTISQRIVSA QSLAEDDV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
TOMM20 Polyclonal antibody specifically detects TOMM20 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TOMM20 | |
Polyclonal | |
Unconjugated | |
TOMM20 | |
1810060K07Rik; BB284719; Gm19268; KIAA0016; MAS20; mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; mKIAA0016; MOM19; outer mitochondrial membrane receptor rTOM20; outer mitochondrial membrane receptor Tom20; TOM20; TOMM20; translocase of outer mitochondrial membrane 20; translocase of outer mitochondrial membrane 20 homolog (yeast); translocase of outer mitochondrial membrane 20 homolog type II | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
67952, 9804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15388, Q9DCC8 | |
TOMM20 | |
Recombinant protein corresponding to Human TOMM20 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only