Learn More
Emma Safety Footwear Tom Safety Shoes
The ideal choice for those who spend hours on their feet such as professionals in Catering, Security, and the Services Industry. There's no toe cap so weight is remarkably reduced, making it an active, very comfortable and versatile shoe that's easy to clean and maintain.
Brand: Emma Safety Footwear MM70954248
Additional Details : Weight : 1.61300kg
Description
- O2-Work shoe without steel toecap
- Full grain leather construction
- GripForce™ Easy Twist
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20347:2012 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
48 |
Laces | |
Tom | |
Leather |