missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Gel Permeation Chromatography (GPC), Fisher Chemical

Catalog Number 10346440
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Melting Point -95°C
CAS 108-88-3
Color Colorless
Boiling Point 111°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Molecular Formula C7H8
MDL Number MFCD00008512
Vapor Pressure 29mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
PubChem CID 1140
ChEBI CHEBI:17578
Formula Weight 92.14g/mol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'