missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene-d{8}, 100% (Isotopic), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 Ea.
2 Ea.
Description
Toluene-d{8} is used to produce d7-4-methylacetophenone by heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Density | 0.94 g/mL |
Flash Point | 10°C (50°F) |
Odor | Aromatic |
Packaging | Prescored Ampule |
Quantity | 10 Ea. |
Melting Point | -93°C |
Boiling Point | 111°C |
UN Number | UN1294 |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'