missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Certified AR for Analysis, Fisher Chemical™
64.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
64.87 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
64.87 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
C7H8, CAS Number-108-88-3, monomethyl benzene, phenylmethane, methylbenzene, methylbenzol, benzene, methyl, methacide, antisal 1a, tolu-sol, toluen, toluol, 2.5L, 111 deg.C, CHEBI:17578, Colorless, 92.14g/mol, YXFVVABEGXRONW-UHFFFAOYSA-N, toluene, -95 deg.C, 8512, 92.141, Amber glass bottle, Liquid
Specifications
Melting Point | -95°C |
Color | Colorless |
Boiling Point | 111°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 29mbar at 20°C |
Viscosity | 0.6 mPaS at 20°C |
Formula Weight | 92.14g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'