missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
20 L
4 x 2.5 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -93°C (literature) |
CAS Max % | 100.0000% |
Linear Formula | C6H5CH3 |
UN Number | UN1294 |
Beilstein | 635760 |
Formula Weight | 92.14 |
Percent Purity | >99.5% |
Grade | ACS Reagent |
Density | 0.865 g/mL at 25 °C(lit.) |
Boiling Point | 110.6°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'