missing translation for 'onlineSavingsMsg'
Learn More

Toluene, 99+%, extra pure, ACROS Organics™

Brand:  Acros Organics 177160050

Code : 50

Additional Details : CAS Number : 108-88-3 Weight : 4.38000kg Transport : UN number : 1294 Chem class : 1 Pack group : II


Quantity 5L
Packaging Plastic drum

 View more versions of this product

Product Code. 10407153

  • £77.30 / 5 L


Estimated Shipment
15-11-2017
Add to basket


Specifications

Chemical Identifiers

Toluene
Extra Pure
99+%
C6H5CH3
MFCD00008512
108-88-3
≥99%
C7H8
92.14
Methylbenzene

Specifications

Toluene
Vapor Pressure: 29mbar at 20°C
Liquid
99+%
C6H5CH3
MFCD00008512
15, 9688
Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Plastic drum
0.003% max.
0.866
4°C
111.0°C
− 95.0°C
108-88-3
Extra Pure
≥99%
C7H8
92.14
05, 280
Methylbenzene
0.8660g/mL
1.4968
Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
0.05% max. (K.F.)
0.6 mPa.s (20°C)
Clear
5L
Safety and Handling

Safety and Handling

Product Identifier
  • Toluene

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

Documentation