missing translation for 'onlineSavingsMsg'
Learn More

Tocris™olve 100, Tocris Bioscience™

Water-soluble emulsion; negative control for reagents in TocrisolveTM 100

Brand:  RD Systems (Bio-Techne) 16841MIL

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 1mL

 View more versions of this product

Product Code. 13429913

  • £114.66 / 1mL

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Water soluble emulsion composed of a 1:4 ratio of soya oil/water that is emulsified with the block co-polymer Pluronic F68. Can be diluted with any aqueous medium. Used to solubilize hydrophobic organic chemicals and water-insoluble lipids such as cannabinoids and vanilloids. The cannabinoid or vanilloid is incorporated with the emulsion as part of a specialized innovative production process. The soya oil forms a coating around the cannabinoid molecule, and the block co-polymer is added to stabilize the emulsion, preventing the lipid droplets from coalescing (merging) in the surrounding water. Also can be used as a negative control for reagents in TocrisolveTM 100; Anandamide, AM 404, VDM 11, ACPA, (R)-(+)-Methanandamide, JWH 133 and Arachidonic acid (Cat. Nos. 1017, 1685, 1686, 1781, 1782, 1783 and 2756 respectively).

Tocrisolve™ 100 Technical Support .

Specifications

Chemical Identifiers

Specifications

1mL
Tocrisolve™ 100
>98%
Videos
Documents