Learn More
Tocris Bioscience™ ICI 174,864
δ selective peptide antagonist
Brand:  Tocris Bioscience™ 08205
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13488033
  • £492.72 / 5mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Selective δ opioid antagonist. Exhibits partial agonist in vitro activity at δ receptors at high concentrations.
Specifications
Specifications
691.87
purity determined by batch