Learn More
Thermo Scientific Chemicals Tobramycin, 97%
Use promo code "21615" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Tobramycin is an aminoglycoside compound derived from kanamycin B and displays antibiotic and bacteriostatic activity against gram-negative bacteria
• After entering the cells, it binds irreversibly to a specific aminoglycoside receptor on the bacterial 30S ribosomal subunit and interferes with the initiation complex between messenger RNA and the 30S subunit, consequently inhibiting the protein synthesis process leading to bacterial cell death
• Tobramycin also can interfere with protein elongation since it can induce misreading of the mRNA template, and cause incorrect amino acids to be incorporated into the growing polypeptide chain
Specifications
Type | Tobramycin |
Infrared Spectrum | Authentic |
Solubility Information | Solubility in water: soluble in cold water. |
Formula Weight | 467.51 |
Percent Purity | 97% |
Water | 8% max. |
Quantity | 1 g |
Chemical Name or Material | Tobramycin |
Safety and Handling
- Tobramycin
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only