Learn More
TNMD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557096
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GIEFDPMLDE RGYCCIYCRR GNRYCRRVCE PLLGYYPYPY CYQGGRVICR V Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
TNMD is a single-pass type II membrane proteinPotential. It belongs to the chondromodulin-1 family and contains 1 BRICHOS domain. TNMD may be an angiogenesis inhibitor.Specifications
TNMD | |
Polyclonal | |
Unconjugated | |
TNMD | |
1110017I01Rik; Bricd4; BRICHOS domain containing 4; ChM1L; Chondromodulin-1-like protein; chondromodulin-IB; chondromodulin-I-like protein; hChM1L; hTeM; mChM1L; mTeM; myodulin; rChM1L; rTeM; teM; Tendin; tenomodulin; TNMD; UNQ771/PRO1565; zgc:172161 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64102 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2S6 | |
TNMD | |
Recombinant protein corresponding to Human TNMD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only