missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNK2 (ACK) Recominant Human Protein
TNK2 (ACK) Recominant Human Protein
Brand: Invitrogen PV4807
Code : 50
Additional Details : Weight : 0.50000kg
Description
TNK2 (ACK) (activated Cdc42-associated kinase) is a non-receptor tyrosine kinase that signals downstream of Cdc42. TNK2 is known to be involved in tumorigenesis and metastasis, particularly in prostate cancer.Specifications
NP_005772.3 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
69.3kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
TNK2 (ACK) | |
TNK2 (ACK) was pre-diluted in enzyme dilution buffer and assayed in 20mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 1mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.02% Triton™ X-100, 200μM ATP, 200μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 110-476 | |
99nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL | |
GST Tag | |
Kinase and inhibitors | |
95% by SDS-PAGE |
0.31mg/mL | |
10188 | |
TNK2 (ACK) Recombinant Human Protein | |
–80°C | |
1 Each | |
TNK2 (ACK) Recombinant Human Protein | |
ACK | |
TNK2 (ACK) Recombinant Human Protein | |
Human | |
Baculovirus | |
Cytoplasmic tyrosine kinase |
For Research Use Only. Not for use in diagnostic procedures.