missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ TNFRSF4 Recombinant Human Protein, hIgG1-Fc.His Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Ala 216) of human TNFRSF4 precursor fused to C-terminal polyhistidine-tagged Fc region of human IgG1

Brand:  Sino Biological™ 10481H03H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15246795

  • £125.97 / 25µg


Estimated Shipment
09-05-2018
Add to basket

Description

Description

  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
22163
>85% by SDS-PAGE
Human
<1 EU/μg
TNFRSF4
Cytokines and Receptors, CD Proteins
TNFRSF4 Recombinant Human Protein
Lyophilized
48.2kd
5 x 5μg
Truncated, Recombinant
Tumor Necrosis Factor Receptor Superfamily, Member 4
Assay Standard
Mammalian
hIgG1-Fc.His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.