missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RII/TNFRSF1B Hamster anti-Mouse, Clone: TR75-89.29, R&D Systems™
Hamster Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | TNF RII/TNFRSF1B |
---|---|
Clone | TR75-89.29 |
Immunogen | E. coli-derived recombinant mouse TNF RII/TNFRSF1B |
Gene ID (Entrez) | 7133 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
TNF RII/TNFRSF1B | |
E. coli-derived recombinant mouse TNF RII/TNFRSF1B | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse TNF RII/TNFRSF1B in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot, Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
TR75-89.29 | |
7133 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Hamster | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
For Research Use Only