missing translation for 'onlineSavingsMsg'
Learn More

TNF alpha Rat anti-Mouse, PE-eFluor 610, Clone: MP6-XT22, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-7321-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15550567

  • £199.23 / 100µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

The MP6-XT22 antibody reacts with mouse tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4(+) T cells. TNF alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane-bound form.
Specifications

Specifications

TNF alpha
MP6-XT22
PE-eFluor 610
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Flow Cytometry (Intracellular Staining)
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Tumor Necrosis Factor alpha
IgG1
RUO
Primary
Murine
Videos