missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF alpha, PE, clone: MP6-XT22, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 12-7321-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The MP6-XT22 antibody reacts with mouse tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4(+) T cells. TNF alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane-bound form.Applications Reported: This MP6-XT22 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This MP6-XT22 antibody is tested by intracellular flow cytometric analysis of stimulated splenocytes. This product has been pre-titrated and tested by intracellular flow cytometric analysis. This can be used at 5μL (0.125μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
TNF alpha | |
Monoclonal | |
5μl/Test | |
PBS with 0.2% BSA, 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P06804 | |
DADB-70P7.1, DIF, TNF-alpha, TNFA, TNFSF2 | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
Flow Cytometry | |
MP6-XT22 | |
PE | |
Tnf | |
Conjugated | |
Tnf | |
IgG1, kappa | |
25 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
21926 |
For Research Use Only.