missing translation for 'onlineSavingsMsg'
Learn More

TNF alpha, clone: 1F3F3D4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-7325-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15278067

  • £143.44 / 500µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Description: The 1F3F3D4 antibody reacts with mouse tumor necrosis factor-alpha (TNFaalpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4(+) T cells.Applications Reported: The 1F3F3D4 antibody has been reported for use in capture of mouse TNF alpha by ELISA and ELISPOT.Applications Tested: The 1F3F3D4 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse TNF alpha (Tumor Necrosis Factor alpha). A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TNF alpha
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P06804
DADB-70P7.1, DIF, TNF-alpha, TNFA, TNFSF2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
ELISA
1F3F3D4
Unconjugated
Tnf
Purified
Tnf
IgM, kappa
500μg
Store at 2-8°C.
Primary
21926
Videos

For Research Use Only.