missing translation for 'onlineSavingsMsg'
Learn More

TNF alpha, PE, clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7349-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209329

  • £84.99 / 50µg


EU Stock 5
Add to basket

Description

Description

The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TN alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.
Specifications

Specifications

TNF alpha
MAb11
PE
Conjugated
Murine
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL; 5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Tumor Necrosis Factor alpha
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.