missing translation for 'onlineSavingsMsg'
Learn More

TNF alpha, Biotin, clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-7349-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15330070

  • £158.21 / 500µg


EU Stock 9
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF-alpha ), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4^+ T cells. TNF-alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF-alpha forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.Applications Reported: The MAb11 antibody has been reported for use in detection of human TNF alpha by ELISA.Applications Tested: The MAb11 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of human TNF-alpha (TNF-a, TNFa) in combination with the purified MAb1 (14-7348) antibody for capture and recombinant human TNFa (14-8329) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TNF alpha
MAb11
Biotin
Conjugated
Murine
500μg
Store at 2-8°C. Do not freeze.
Monoclonal
ELISPOT Detection Antibody
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Tumor Necrosis Factor alpha
IgG1
RUO
Primary
Human
Videos