missing translation for 'onlineSavingsMsg'
Learn More

TNF alpha, Alexa Fluor 700, clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-7349-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510377

  • £74.67 / 25 tests


Estimated Shipment date
from Supplier 26-07-2018
Add to basket

Description

Description

Description: The MAb11 antibody reacts with human tumor necrosis factor-alpha (TNF alpha), a 17kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4+ T cells. TN alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26kDa membrane bound form. The MAb11 antibody is a neutralizing antibody.Applications Reported: This MAb11 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This MAb11 antibody has been pre-titrated and tested byintracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TNF alpha
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01375
DADB-70P7.1, DIF, TNF-alpha, TNFA, TNFSF2
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MAb11
Alexa Fluor 700
TNF
Conjugated
TNF
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
7124
Videos

For Research Use Only.