missing translation for 'onlineSavingsMsg'
Learn More

TNF alpha Mouse anti-Human, Alexa Fluor® 488, Clone: MAb11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-7349-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15590247

  • £71.75 / 25 tests

Estimated Shipment
03-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
Specifications

Specifications

TNF alpha
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01375
DADB-70P7.1, DIF, TNF-alpha, TNFA, TNFSF2
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MAb11
Alexa Fluor™ 488
TNF
Liquid
TNF
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
7124
Videos
Documents

For Research Use Only.