missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TNF alpha ELISA Kit, Porcine
TNF alpha ELISA Kit, Porcine
Brand: Thermo Scientific™ EP2TNFA2
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Assay TNFa (porcine tumor necrosis factor alpha) levels in pig serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Porcine TNF alpha ELISA Kit.Features of the Porcine TNF alpha ELISA Kit:
This porcine TNF alpha ELISA kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of tumor necrosis factor alpha (TNF-alpha, TNFα) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kit contains precoated 96-well strip plates and lot-optimized reagents.
Specifications
31.3 to 2000pg/mL | |
TNF alpha ELISA Kit, 192 Assays, 3.1 to 2000pg/mL Test Range, 4 hr. Test Time, <0.5pg/mL Sensitivity, Cell Culture Supernatants, Plasma, Serum Sample, Swine/Porcine Species, Colorimetric Detection Method, TNFα Gene, Used for Protein Assays and Analysis | |
192 | |
Colorimetric ELISA (TMB substrate) with Plates | |
Cytokines and receptors | |
No significant cross-reaction with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12p70, IL-12p40, IL-13, IL-15, IL-16, IL-17, IL-18, Eotaxin, GCSF, GMCSF, GROα, GROβ, IFNγ, IFNα, MCP-1, MCP-2, MCP-3, MCP-4, VEGF, MIP-1α, MI | |
192 Assay | |
Protein assays and analysis | |
Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s) and stop solution | |
Dependent upon sample type and level | |
For research use only (RUO) | |
50 μL | |
Natural and recombinant porcine TNFα± | |
4 hr. |
<5pg/mL | |
TNF-a (TNF) | |
Colorimetric | |
TNF alpha ELISA Kit | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
TMB | |
Porcine, Swine | |
4 hr. | |
Absorbance at 450nm minus absorbance at 550nm | |
192 Assays | |
Immunology | |
2°C to 8°C | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.