Learn More
TMPH hydrochloride, Tocris Bioscience™
Neuronal nicotinic receptor antagonist
Brand:  Tocris (Bio-Techne) 243810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13461204
  • £128.25 / 10mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Potent non-competitive antagonist of neuronal nicotinic ACh receptors (nAChRs). Produces long-lasting inhibition of neuronal nAChRs formed by the combination of the most abundant α and β subunits (i.e. α3, α4 and β2, β4 respectively). Displays little inhibition of muscle-type (α1β1γδ) or α7 receptors.
Specifications
Chemical Identifiers
849461-91-2
305.887
tmph hydrochloride, 2,2,6,6-tetramethylpiperidin-4-yl heptanoate hydrochloride, 2,2,6,6-tetramethyl-4-piperidyl heptanoate hydrochloride, heptanoic acid, 2,2,6,6-tetramethyl-4-piperidinyl ester, hydrochloride, tmph hydrochloride, solid, product is pure based on chn, nmr and ms results
(2,2,6,6-tetramethylpiperidin-4-yl) heptanoate;hydrochloride
C16H32ClNO2
XIDDVJIJIFWGIX-UHFFFAOYSA-N
16079014
CCCCCCC(=O)OC1CC(NC(C1)(C)C)(C)C.Cl
Specifications
TMPH hydrochloride
849461-91-2
tmph hydrochloride, 2,2,6,6-tetramethylpiperidin-4-yl heptanoate hydrochloride, 2,2,6,6-tetramethyl-4-piperidyl heptanoate hydrochloride, heptanoic acid, 2,2,6,6-tetramethyl-4-piperidinyl ester, hydrochloride, tmph hydrochloride, solid, product is pure based on chn, nmr and ms results
CCCCCCC(=O)OC1CC(NC(C1)(C)C)(C)C.Cl
305.887
305.88
10mg
C16H32ClNO2
XIDDVJIJIFWGIX-UHFFFAOYSA-N
(2,2,6,6-tetramethylpiperidin-4-yl) heptanoate;hydrochloride
16079014
>98%
Videos