Learn More
TMEM8A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562365
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TCAYVFQPEL LVTRVVEISI MEPDVPLPQT LLSHPSYLKV FVPDYTRELL LELRDCVSNG SLGCPVRLTV GPVTLPSNFQ KVL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
TMEM8A Polyclonal antibody specifically detects TMEM8A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
TMEM8A | |
Polyclonal | |
Unconjugated | |
TMEM8A | |
five-span transmembrane protein M83; GPI processing phospholipase A2; GPI-PLA2; M83; PGAP6; Post-GPI attachment to proteins factor 6; Protein M83; TMEM6; TMEM8; TMEM8A; Transmembrane protein 6; Transmembrane protein 8; transmembrane protein 8 (five membrane-spanning domains); transmembrane protein 8A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCN3 | |
TMEM8A | |
Recombinant protein corresponding to Human TMEM8A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only