missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM221 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559636
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RGLHELSPPS FEDDLARPAE VSKASPRAQP QQGIHRRTPY STCP Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 55%.
TMEM221 is a protein coding gene.Specifications
TMEM221 | |
Polyclonal | |
Unconjugated | |
TMEM221 | |
Putative transmembrane protein ENSP00000342162; TMEM221; Transmembrane protein 221 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100130519 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NGB7 | |
TMEM221 | |
Recombinant protein corresponding to Human TMEM221 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only