missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM141 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565603
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MVNLGLSRVDD AVAAKHPGLG EYAACQSH Highest antigen sequence identity to the following orthologs - mouse 90%, rat 93%.
TMEM141 Polyclonal antibody specifically detects TMEM141 in Human samples. It is validated for Immunocytochemistry.Specifications
TMEM141 | |
Polyclonal | |
Unconjugated | |
TMEM141 | |
1110065P14Rik; AI663999; D2Ertd217e; RGD1561492; Tmem141; Transmembrane protein 141 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85014 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96I45 | |
TMEM141 | |
Recombinant Human TMEM141 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only