missing translation for 'onlineSavingsMsg'
Learn More

TLR4/MD-2 Complex, clone: MTS510, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9924-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238247

  • £143.16 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The MTS510 monoclonal antibody reacts with the mouse Toll-like receptor 4 (TLR4)/MD-2 complex. At least ten members of the Toll family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR4 physically associates with MD-2, and together with CD14, this complex is responsible for LPS recognition and signaling. In the mouse, TLR4 is expressed by thioglycolate-elicited peritoneal macrophages. Incubation of peritoneal macrophages with LPS results in down regulation of surface TLR4/MD-2. The TLR4 gene is defective in C3H/HeJ and C57BL/10ScCr mice, both of which have been well characterized as hyporesponders to LPS. The MTS510 monoclonal antibody co-immunoprecipitates MD-2 (∽30 kDa) and TLR4 (∽100 kDa), and preferentially reacts with TLR4 that is associated with MD-2. In comparison, binding of the UT41 monoclonal antibody occurs with and without formation of the TLR4/MD-2 complex. Please contact eBioscience Technical Support for further information.Applications Reported: This MTS510 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunohistology staining of frozen tissue sections and inhibition of LPS-induced cytokine production. (Please use Functional Grade purified MTS510, cat. 16-9924, in functional assays.).Applications Tested: The MTS510 antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TLR4/MD2
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9QUK6, Q9JHF9
TLR4/ MD-2, TLR-4/ MD2, toll-like receptor MD2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Immunoprecipitation
MTS510
Unconjugated
Tlr4
Purified
Ly96, Tlr4
IgG2a, kappa
100μg
Store at 2-8°C.
Primary
17087, 21898
Videos

For Research Use Only.