missing translation for 'onlineSavingsMsg'
Learn More

Grace Davison Discovery Scienc TLC tank 12cm x 11.5cm x 8.6cm developing chamber for 10cm x 10cm TLC plates

Brand:  Grace Davison Discovery Scienc 2108345

TLC tank 12cm x 11.5cm x 8.6cm developing chamber for 10cm x 10cm TLC plates

Code : 17

Additional Details : Weight : 1.00000kg

Product Code. 13015803

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.