Learn More
TL1A Monoclonal Antibody (Tandys1a), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-7911-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The Tandys1a monoclonal antibody reacts with human and mouse TL1A (TNFSF15). TL1A is a member of the TNF ligand family and is expressed by endothelial cells, tissue macrophages, monocyte-derived dendritic cells, plasma cells and lamina propria-derived lymphocytes. Among peripheral blood cells, TL1A has been shown to be expressed predomitly by CD4+CCR9+ lymphocytes. Its expression has been shown to be induced by TNF alpha, IL-1 alpha and IFN gamma. TL1A binds DR3, which has been shown to be specifically upregulated in Th17 cells among CD4+ T cells. TL1A-deficient dendritic cells are weak in their ability to support Th17 cells in vitro and show decreased severity of EAE. In vivo, TL1A has been shown to be upregulated in tissue lymphocytes of Crohn's Disease and ulcerative colitis patients. Applications Reported: This Tandys1a antibody has been reported for use in flow cytometric analysis, and intracellular staining followed by flow cytometric analysis. Applications Tested: This Tandys1a antibody has been tested by flow cytometric analysis of normal human peripheral blood cells or mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
TL1A Monoclonal antibody specifically detects TL1A in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
TL1A | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
O95150, Q5UBV8 | |
TNFSF15 | |
Human TL1A recombinant ECD | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
Tandys1a | |
PerCP-eFluor 710 | |
TNFSF15 | |
bM20K13.3 (tumor necrosis factor (ligand) superfamily, member 15); MGC129934; MGC129935; TL1; TL1A; TNF ligand-related molecule 1; TNF superfamily ligand TL1A; TNFSF15; TNLG1B; tumor necrosis factor (ligand) superfamily member 15; tumor necrosis factor (ligand) superfamily, member 15; tumor necrosis factor ligand 1B; tumor necrosis factor ligand superfamily member 15; Tumor necrosis factor ligand superfamily member 15, membrane form; Tumor necrosis factor ligand superfamily member 15, secreted form; tumor necrosis factor superfamily member 15; vascular endothelial cell growth inhibitor; vascular endothelial growth inhibitor; vascular endothelial growth inhibitor-192A; Vegi; VEGI192A | |
Mouse | |
Affinity chromatography | |
RUO | |
326623, 9966 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
For Research Use Only.