Learn More
Tocris Bioscience™ Tizanidine hydrochloride
α2 agonist
POA
Specifications
Purity >99%
View More Specs
Description
Description
α2-adrenergic receptor agonist. Antinociceptive upon epidural administration in rats (IC50 = 48 nM). Also binds to imidazoline receptor.
Specifications
Specifications
>99%