Learn More
Tocris Bioscience™ Tizanidine hydrochloride
α2 agonist
Brand:  Tocris Bioscience™ 360950
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13453284
  • £170.52 / 50mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
α2-adrenergic receptor agonist. Antinociceptive upon epidural administration in rats (IC50 = 48 nM). Also binds to imidazoline receptor.
Specifications
Specifications
Tizanidine hydrochloride
>99
50mg
Tizanidine hydrochloride
290.17