Learn More
Tocris Bioscience™ Tizanidine hydrochloride
α2 agonist
Brand:  Tocris Bioscience™ 360910
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443284
  • £52.65 / 10mg
Estimated Shipment
08-07-2019
Description
Description
α2-adrenergic receptor agonist. Antinociceptive upon epidural administration in rats (IC50 = 48 nM). Also binds to imidazoline receptor.
Specifications
Specifications
Tizanidine hydrochloride
>99
10mg
Tizanidine hydrochloride
290.17