missing translation for 'onlineSavingsMsg'
Learn More

Titanium Thinfoil, 0.006mm (0.0002in) thick, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog Number 11318717
74.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]

Form: Approx. 0.02g/25 x 25mm; Not certified pinhole free

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray foil
Assay Percent Range 99.6% (Metals basis)
Quantity 25 x 25 mm
Odor Odorless
Chemical Name or Material Titanium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'