missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Ea.
Description
As intermediates and as salt. Used in coating, vesicant and powder metallurgy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 Ea. |
CAS | 7440-32-6 |
Odor | Odorless |
Molecular Formula | Ti |
MDL Number | MFCD00011264,MFCD00151301 |
Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
Solubility Information | Insoluble in water. |
InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
SMILES | [Ti] |
IUPAC Name | titanium |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'