missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium powder, <20 micron, 93%, packaged in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Gray |
Appearance | Dark Gray powder |
Assay Percent Range | 93% |
Quantity | 500 g |
Odor | Odorless |
Form | Powder |
Chemical Name or Material | Titanium |
Safety and Handling
Product Identifier
- Titanium powder
- Danger
- Flammable solid Category 1
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P370+P378-In case of fire: Use special powder for metal fires to extinguish.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'