missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, rutile, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11316948
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1317-80-2
O2Ti
79.87
MFCD00011269,MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1317-80-2
O2Ti
79.87
MFCD00011269,MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O

Titanium(IV) oxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2500°C to 3000°C
Melting Point 1830°C to 1850°C
Color White
Physical Form Powder
Quantity 50 g
Merck Index 14,9472
Solubility Information Soluble in hot concentrated sulfuric acid,in hydrofluoric acid and alkali. Insoluble in water.
Formula Weight 79.9
Percent Purity 99.99%
Odor Odorless
Density 4.23 g/mL
Assay (metals basis)
Chemical Name or Material Titanium(IV) oxide, Rutile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'