missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, 98+%, anatase powder, Thermo Scientific Chemicals

Catalog Number p-221751
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
13463-67-7
O2Ti
79.87
MFCD00011269 MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O
This item is not returnable. View return policy
13463-67-7
O2Ti
79.87
MFCD00011269 MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2900.0°C
Melting Point 1855.0°C
Color White
pH 7.3 to 8.3 (aq. suspension)
Physical Form Powder
Quantity 100 g
Assay Percent Range 98% min
Linear Formula TiO2
Merck Index 15, 9628
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
Formula Weight 79.88
Percent Purity ≥98%
Packaging Glass bottle
Specific Gravity 4.23
Density 4.2300g/mL
Chemical Name or Material Titanium(IV) oxide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'