missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) n-butoxide, 99+%, Thermo Scientific Chemicals

Catalog Number 15472957
Click to view available options
Quantity:
10 g
100 g
500 g
5593-70-4
C16H40O4Ti
344.359
MFCD00009433
FPCJKVGGYOAWIZ-UHFFFAOYSA-N
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
25203927
butan-1-ol;titanium
CCCCO.CCCCO.CCCCO.CCCCO.[Ti]
5593-70-4
C16H40O4Ti
344.359
MFCD00009433
FPCJKVGGYOAWIZ-UHFFFAOYSA-N
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
25203927
butan-1-ol;titanium
CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Titanium(IV) n-butoxide is used in the preparation of nanosized titania powders in the anatase form and ferroelectric bismuth titanate thin films. It is also used as intermediates, paint additives, coating additives, processing aids and as process regulators. It is involved in the preparation of nanocrystalline titanium dioxide powders at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 310°C to 314°C
Melting Point -55°C
Color Yellow
Physical Form Liquid
Quantity 100 g
Linear Formula Ti[O(CH2)3CH3]4
UN Number UN2924
Beilstein 4148236
Flash Point 41°C (170°F)
Solubility Information Miscible with aliphatic,aromatic,chlorinated and oxygenated solvents.
Formula Weight 340.36
Percent Purity ≥99%
Odor Alcohol-like
Refractive Index 1.49
Sensitivity Moisture sensitive
Density 0.998 g/mL
Chemical Name or Material Titanium (IV) n-butoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) n-butoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Butyl titanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'