missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals

Catalog Number 15432957
Click to view available options
Quantity:
10 g
100 g
500 g
4 x 500 g
546-68-9
C12H28O4Ti
284.219
MFCD00008871
VXUYXOFXAQZZMF-UHFFFAOYSA-N
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
11026
propan-2-olate;titanium(4+)
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
546-68-9
C12H28O4Ti
284.219
MFCD00008871
VXUYXOFXAQZZMF-UHFFFAOYSA-N
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
11026
propan-2-olate;titanium(4+)
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids.Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 232°C
Melting Point 16°C to 20°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 100 g
Linear Formula Ti[OCH(CH3)2]4
UN Number UN2413
Beilstein 3679474
Flash Point 46°C (115°F)
Merck Index 14,9480
Solubility Information Soluble in anhydrous ethanol, ether, benzene and chloroform; Decomposes in water
Formula Weight 284.23
Percent Purity ≥97%
Refractive Index 1.464
Sensitivity Moisture sensitive
Density 0.955 g/mL
Chemical Name or Material Titanium (IV) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'