Learn More
Titanium(IV) isopropoxide, 95%, Thermo Scientific Chemicals
Description
Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.
Solubility
Soluble in anhydrous ethanol, ether, benzene and chloroform.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids. It reacts with water to produce titanium dioxide.
Specifications
Melting Point | 16°C to 20°C |
Density | 0.955 |
Boiling Point | 232°C |
Flash Point | 46°C (115°F) |
Linear Formula | Ti[OCH(CH3)2]4 |
Refractive Index | 1.464 |
Quantity | 100 g |
UN Number | UN2413 |
Beilstein | 3679474 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only