missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) iodide, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
85.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Titanium(IV) iodide, anhydrous is used in the CVD of titanium nitride films with optimized characteristics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 150°C |
Physical Form | Lump |
Quantity | 10 g |
UN Number | UN3260 |
Solubility Information | Soluble in dichloromethane,chloroform,carbon disulfide. |
Formula Weight | 555.52 |
Percent Purity | 99.9% |
Odor | Acrid |
Sensitivity | Moisture sensitive |
Density | 4.30 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'